Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine hydrochloride, MP Biomedicals™

Catalog No. ICN19067525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19067525 25 g
ICN19067510 10 g
ICN19067580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19067525 Supplier MP Biomedicals, Inc Supplier No. 0219067525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8063-24-9
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.91
MDL Number MFCD00069039
InChI Key MKLTXAHQKDVBLY-UHFFFAOYSA-N
Synonym Euflavin, Trypaflavine
IUPAC Name 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
SMILES Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Specifications

Assay Percent Range 0.991
Formula Weight 245.8
Color Brown/Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Acriflavine hydrochloride, Intercalating dye, antiseptic, analysis of cellular DNA and protein
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.