Learn More
Cytiva Acrodisc Syringe Filter with Supor Membrane - 0.45 µm, 25 mm modified acrylic housing (1000/pkg)
Supplier:  Cytiva 4508
Encompass
Non-sterile syringe filters with a Supor membrane in modified acrylic housing can be used for the filtration of general aqueous solutions. They are available in a 13 mm, 25 mm and 32 mm diameters, and a range of pore sizes from 0.1 μm to 5 μm are available to suit different application requirements.Supor polyethersulfone (PES) membrane has been optimized for biological, pharmaceutical, and sterilizing filtration requirements. It is a low protein binding membrane making it an excellent choice when minute amounts of sample need to be recovered. The membrane has extensive drug compatibility and moderate chemical resistance and compatibility.Fast filtration with superior flow rates and high throughputsLow protein binding and extensive drug compatibility for critical applicationsLow extractablesHigh consistencyHydrophilic, for the filtration of aqueous solutionsFor research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3058