Learn More
Cytiva (Formerly Pall Lab) Acro 50 Vent Devices with Emflon II Membrane - Hose barb, no vent, 0.2 m 100% integrity tested during manufacturing (100/pkg)
Catalog No. 502063396 Shop All Cytiva Products
Encompass
50 mm Filters with Superior Performance for Demanding ApplicationsReusable and autoclavablePTFE membrane and polypropylene housing have broad chemical resistanceLarge surface area provides greater throughput and high air flow rateAvailable with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3396 Supplier Cytiva Supplier No. A50V002P2NV
50 mm Filters with Superior Performance for Demanding ApplicationsReusable and autoclavablePTFE membrane and polypropylene housing have broad chemical resistanceLarge surface area provides greater throughput and high air flow rateAvailable with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.