Learn More
Cytiva (Formerly Pall Lab) Acro Vent Devices with PTFE Membrane - 0.2 m, 1/8 in. MNPT (18/pkg)
Supplier:  Cytiva 4400
Encompass_Preferred
50 mm Filters with Superior Performance for Demanding ApplicationsReusable and autoclavablePTFE membrane and polypropylene housing have broad chemical resistanceLarge surface area provides greater throughput and high air flow rateAvailable with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3041
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.