Learn More
Cytiva Acro 50 Vent Devices with Emflon II Membrane - Hose barb with vent, 0.2 µm (3/pkg)
Catalog No. NC1673821 Shop All Cytiva Products
Encompass
50 mm Filters with Superior Performance for Demanding Applications•Reusable and autoclavable•PTFE membrane and polypropylene housing have broad chemical resistance•Large surface area provides greater throughput and high air flow rate•Available with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1673821 Supplier Cytiva Supplier No. A50V002P2
50 mm Filters with Superior Performance for Demanding Applications•Reusable and autoclavable•PTFE membrane and polypropylene housing have broad chemical resistance•Large surface area provides greater throughput and high air flow rate•Available with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.