Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filters with Nylon Membrane - 0.45 m, 13mm, minspike outlet (1000/pkg)
Catalog No. 502063065 Shop All Cytiva Products
Encompass
Syringe filters with nylon membrane are ideal for the filtration of aqueous and solvent-based sample prep for HPLC (0.45 m) and UHPLC (0.2 m) applications. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime.Nylon syringe filters show excellent chemical compatibility with esters, bases and alcohols. Nylon syringe filters are not recommended for use with strongly acidic fluids.Designed for the filtration of sample volumes up to 10 mL. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results.Pall 13 mm syringe filters are available with a male slip luer outlet or a proprietary minispike outlet. The minispike allows for easy handling with a HPLC vial and offers a reduced hold-up volume. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3065 Supplier Cytiva Supplier No. 4546
Add to Cart
Add to Cart
Syringe filters with nylon membrane are ideal for the filtration of aqueous and solvent-based sample prep for HPLC (0.45 m) and UHPLC (0.2 m) applications. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime.Nylon syringe filters show excellent chemical compatibility with esters, bases and alcohols. Nylon syringe filters are not recommended for use with strongly acidic fluids.Designed for the filtration of sample volumes up to 10 mL. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results.Pall 13 mm syringe filters are available with a male slip luer outlet or a proprietary minispike outlet. The minispike allows for easy handling with a HPLC vial and offers a reduced hold-up volume. This product is not approved for use in medical, clinical, surgical or other patient applications.