Learn More
Cytiva (Formerly Pall Lab) Acrodisc™ One Syringe Filters with wwPTFE membrane - 0.45 m AutoPack tubes (25/pkg, 200/cs)
Catalog No. 502063431 Shop All Cytiva Products
Encompass
wwPTFE membrane syringe filters are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 m) and UHPLC (0.2 m) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL and features a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems and supplied in ready to use autopack tubes that can be easily loaded into systems.The syringe filters are HPLC certified for low levels of UV-absorbing extractables, and are low in protein and API binding. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3431 Supplier Cytiva Supplier No. AP4914
wwPTFE membrane syringe filters are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 m) and UHPLC (0.2 m) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Acrodisc PSF Syringe Filters have a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL and features a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems and supplied in ready to use autopack tubes that can be easily loaded into systems.The syringe filters are HPLC certified for low levels of UV-absorbing extractables, and are low in protein and API binding. This product is not approved for use in medical, clinical, surgical or other patient applications.