Learn More
Cytiva (Formerly Pall Lab) Acrodisc™ One Syringe Filters with wwPTFE membrane - 0.45 m(1000/pkg)
Catalog No. 502063432 Shop All Cytiva Products
Encompass
wwPTFE membrane syringe filters are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 m) and UHPLC (0.2 m) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL and features a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems.The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results. The syringe filters are low in protein and API binding which make them ideal for drug studies. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3432 Supplier Cytiva Supplier No. AP4915
wwPTFE membrane syringe filters are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 m) and UHPLC (0.2 m) sample prep. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL and features a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems.The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results. The syringe filters are low in protein and API binding which make them ideal for drug studies. This product is not approved for use in medical, clinical, surgical or other patient applications.