Learn More
Cytiva Acrodisc Syringe Filter with PTFE Membrane -1 µm, 25 mm (1000/pkg)
Supplier:  Cytiva 4503
Encompass_Preferred
Acrodisc Syringe Filters with PTFE Membrane - 1 µm, Standard 25mm.Syringe filters with PTFE membrane are ideal for the filtration of organic solvents and harsh solvents in analytical sample preparation applications. PTFE is a hydrophobic membrane and is not suitable for the filtration of aqueous solutions, however these syringe filters are ideal for use in venting and gas filtration applications. 25 mm Standard Acrodisc Syringe Filters are ideal for sample volumes of ≤ 100 mL and feature an effective filtration area of 2.8 cm2. They feature a polypropylene housing that is chemically resistant to harsh solvents and low in extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3056
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.