Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filters with Supor Membrane, Sterile - 0.2 m, 32 mm (50/pkg)
Catalog No. 502063074 Shop All Cytiva Products
Encompass
The syringe filter is the laboratory workhorse that protects sample or culture quality and sterility.Acrodisc syringe filters with Supor membrane have superior performance in a variety of sterile laboratory filtration applications. The inherently hydrophilic Supor (polyethersulfone) membrane is low protein binding and provides fast filtration and high throughput.The sterile Acrodisc syringe filters are individually packaged in blister packs and sterilized using gamma irradiation to eliminate potential or harmful extractables common with other sterilization methods such as EtO.Total throughput (capacity) of a filter depends on the characteristics of the fluid being filtered, such as the extent of microbial contamination, viscosity, and particulate load. For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3074 Supplier Cytiva Supplier No. 4652
The syringe filter is the laboratory workhorse that protects sample or culture quality and sterility.Acrodisc syringe filters with Supor membrane have superior performance in a variety of sterile laboratory filtration applications. The inherently hydrophilic Supor (polyethersulfone) membrane is low protein binding and provides fast filtration and high throughput.The sterile Acrodisc syringe filters are individually packaged in blister packs and sterilized using gamma irradiation to eliminate potential or harmful extractables common with other sterilization methods such as EtO.Total throughput (capacity) of a filter depends on the characteristics of the fluid being filtered, such as the extent of microbial contamination, viscosity, and particulate load. For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.