Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filters with Supor Membrane, Sterile - 0.8 m, 25 mm (50/pkg)
Catalog No. 502063073 Shop All Cytiva Products
Encompass
The inherently hydrophilic Supor (polyethersulfone) membrane is low protein binding and provides fast filtration and high throughput.The sterile Acrodisc syringe filters are individually packaged in blister packs and sterilized using gamma irradiation to eliminate potential or harmful extractables common with other sterilization methods such as EtO.The 0.8 m pore size allows for the prefiltration and clarification of small volumes of buffers, culture media and biological solutions. The 25 mm diameter device is ideal for filtering sample volumes up to 100 mL*.* Total throughput (capacity) of a filter depends on the characteristics of the fluid being filtered, such as the extent of microbial contamination, viscosity, and particulate load. For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3073 Supplier Cytiva Supplier No. 4618
Add to Cart
Add to Cart
The inherently hydrophilic Supor (polyethersulfone) membrane is low protein binding and provides fast filtration and high throughput.The sterile Acrodisc syringe filters are individually packaged in blister packs and sterilized using gamma irradiation to eliminate potential or harmful extractables common with other sterilization methods such as EtO.The 0.8 m pore size allows for the prefiltration and clarification of small volumes of buffers, culture media and biological solutions. The 25 mm diameter device is ideal for filtering sample volumes up to 100 mL*.* Total throughput (capacity) of a filter depends on the characteristics of the fluid being filtered, such as the extent of microbial contamination, viscosity, and particulate load. For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.