Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration - 1 mL, Omega 100K MWCO (5/pkg)
Supplier:  Cytiva 8166
Encompass_Preferred
Provides rapid, efficient separation of biomolecules- Ultrafiltration is the preferred method for biolomecule purification, fractionation, and concentration as well as for buffer exchange and desalting of samples.Omega™ membrane provides high recovery and typically results in > 90% recovery of target biomolecules- For proteins, it is recommended that a molecular weight cut off of 3-6 times smaller than the molecular weight of the solute being retained be selected. New well design results in faster, more uniform filtration rates across the plate with reduced hold-up volume for maximum sample recoveryIntrinsic plate and membrane properties prevent target molecules from binding to the plate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.