Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance - 2 mL, 3.0 m Glass Fiber/0.2 m Supor membrane (5/pkg)
Supplier:  Cytiva 8275
Encompass_Preferred
Prefilter Provides Efficient Clarification of Highly Particulated Samples-Integrated prefilter yields consistent filtration of samples with high levels of gross particulate-New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume-Innovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination-Manufactured with biologically inert materials that allow clarification of most types of lysates without loss of target molecules- Ideal for the removal of bacterial and cellular debris prior to biomolecule purification This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3159
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.