Learn More
Waters Corp Acrodisc, Syringe Filter, PTFE, 25 mm, 0.45 µm, Non polar, 200/case
Supplier:  Waters Corp WAT200501
Encompass_Preferred
This PTFE membrane is a good choice for use with aggresive organic solvents
Catalog No. 50-786-048
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.