Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filters with Glass Fiber - 1 m (nominal) (50/pkg 200/cs)
Catalog No. 501959332 Shop All Cytiva Products
Encompass
Acrodisc Syringe Filters with Glass Fiber, 1 m (nominal).Syringe filters are used in analytical chemistry applications to remove particulate contamination prior to analysis. Filtering a sample prior to sample injection into analytical instrumentation will help protect sensitive parts of the instrument from particulate build-up and reduce system downtime. Acrodisc 25 mm syringe filters with glass fiber maximize throughput prefiltration of particulate-laden samples. They can be used alone or in series with another Acrodisc syringe filter. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 501959332 Supplier Cytiva Supplier No. 4523
Acrodisc Syringe Filters with Glass Fiber, 1 m (nominal).Syringe filters are used in analytical chemistry applications to remove particulate contamination prior to analysis. Filtering a sample prior to sample injection into analytical instrumentation will help protect sensitive parts of the instrument from particulate build-up and reduce system downtime. Acrodisc 25 mm syringe filters with glass fiber maximize throughput prefiltration of particulate-laden samples. They can be used alone or in series with another Acrodisc syringe filter. This product is not approved for use in medical, clinical, surgical or other patient applications.