Learn More
Andwin Scientific ACRODISK CR 25MM 1.0 UM 50/PK
Supplier: Andwin Scientific 4226
The 1.0 m Acrodisc syringe filter with hydrophobic PTFE membrane is suitable with a wide range of chemicals and temperature. The PTFE membrane is also HPLC certified for low levels of UV-absorbing extractables. This product is sold in a pack of 50 filters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.