Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acrolein diethyl acetal

Catalog No. A24001100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24001100G 100 g
A2400125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24001100G Supplier Sigma Aldrich Supplier No. A24001100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrolein diethyl acetal is widely used to carry out chemoselective Heck arylation to synthesize either 3-arylpropanoate esters or cinnamaldehyde derivatives.It can also be used as one of the precursors to synthesize natural products like ()-(Z)-Deoxypukalide, ()-Laulimalide, botryodiplodin, neolaulimalide and isolaulimalide.

Specifications

CAS 3054-95-3
Density 0.854 g/mL (at 25°C (literature))
Boiling Point 125°C (lit.)
Molecular Formula C7H14O2
Refractive Index n20/D 1.398 (literature)
Linear Formula CH2=CHCH(OCH2CH3)2
MDL Number MFCD00009239
Quantity 100 g
Synonym 3,3-Diethoxy-1-propene
Molecular Weight (g/mol) 130.18
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.