Learn More
Description
Evaluate analytical performance at every stage of sample processing. Intended for use as an independent panel for analytical evaluation of BCR-ABL test methods. For research use only.
Full-process panel:
Ensures commutability by using cells to mimic patient specimen and therefore can be processed the same way as your clinical samples.
Aligned with the WHO standard:
Traceable to the World Health Organization's first International genetic Reference Panel for quantitation of BCR-ABL translocation by RQ-PCR, NIBSC Code: 09/138
Sensitive even at MR* 4.5:
The AcroMetrix BCR-ABL Panel offers a dynamic range from 10% to 0.0032%, allowing your lab to analytically validate an extended reportable range for your assay down to the lowest level of molecular response (MR).
FAQs
Specifications
Specifications
| Content And Storage | All vials should be stored immediately at -20°C upon receipt |
| Description | AcroMetrix BCR-ABL Panel (RUO – Not for Clinical Use) |
| Type | BCR-ABL Panel |
| Packaging | One box of controls contains five panel members Panels A-E |
| Quantity | 5 Vials |
| Target Gene | BCR-ABL |
The AcroMetrix BCR- ABL Panel is not CE marked nor has 510(k) clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.