Learn More
Thermo Scientific™ AcroMetrix™ EBV Plasma Controls
GSA_VA
Catalog No. 23279148
Change view
Click to view available options
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-148 Low
23-279-150 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-279-148 Supplier Thermo Scientific™ Supplier No. 961230
Only null left
Add to Cart
Add to Cart
Assess the performance of molecular diagnostic test procedures for EBV with the Thermo Scientific™ AcroMetrix™ EBV Plasma Controls, which are for In Vitro Diagnostic Use.
Assess the analytical performance of molecular diagnostic test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific™ AcroMetrix™ EBV Plasma Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix EBV Low Plasma Control
Detectable Analytes EBV
Type EBV Control
Level Low
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL