Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix EDTA Plasma Dilution Matrix
GSA_VA

Catalog No. 23279170 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
45 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-279-170 45 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-279-170 Supplier Thermo Scientific™ Supplier No. S2284
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepare specimens for testing in plasma-based nucleic acid assays with the Thermo Scientific™ AcroMetrix™ EDTA Plasma Dilution Matrix, which is for Research Use Only.

Prepare specimens for testing in plasma-based nucleic acid assays with the Thermo Scientific™ AcroMetrix™ EDTA Plasma Dilution Matrix (RUO). This matrix is a human plasma matrix shown to be non-reactive for blood borne pathogens, viral nucleic acids and serological markers. It is not intended for use as a substitute for the integral diluents, standards or controls provided by in vitro diagnostic kit manufacturers. The matrix is for Research Use Only and is not for use in diagnostic procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Dilution Matrix
Packaging 45mL
Quantity 45 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.