Learn More
Thermo Scientific™ AcroMetrix™ EV Controls (RUO)
Catalog No. 23279168 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix EV Low Control
AcroMetrix EV Negative Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-279-168 AcroMetrix EV Low Control
23-279-167 AcroMetrix EV Negative Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-279-168 Supplier Thermo Scientific™ Supplier No. 950500
Add to Cart
Add to Cart
Assess the performance of molecular test procedures for EV with the Thermo Scientific™ AcroMetrix™ EV Controls, which are for Research Use Only.
Assess molecular test procedures, including the DNA extraction step, for the qualitative and quantitative determination of enterovirus RNA with the Thermo Scientific™ AcroMetrix™ EV Controls (RUO). The controls are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix EV Low Control
Type EV Control
Level Low
Packaging 5 x 0.3mL
Quantity 5 x 0.3 mL