Learn More
Thermo Scientific™ AcroMetrix™ HBV Controls
GSA_VA
Catalog No. 23279122 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
High
Low
Mid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-122 High
23-279-120 Low
23-279-121 Mid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-122 Supplier Thermo Scientific™ Supplier No. 965003
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid tests for hepatitis B viral DNA (HBV DNA) with these controls, which are for In Vitro Diagnostic Use.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of hepatitis B viral DNA (HBV DNA) with the Thermo Scientific™ AcroMetrix™ HBV Controls. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HBV High Control
Detectable Analytes HBV
Type HBV Control
Level High
Packaging 5 x 1mL
Quantity 5 x 1 mL