Learn More
Thermo Scientific™ AcroMetrix™ HCV Controls
GSA_VA
Catalog No. 23279127 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
High
Low
Mid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-127 High
23-279-125 Low
23-279-126 Mid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-127 Supplier Thermo Scientific™ Supplier No. 963003
Add to Cart
Add to Cart
Identify random or systematic errors in nucleic acid test procedures for hepatitis C viral RNA (HCV RNA) with these controls, which are for In Vitro Diagnostic Use.
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls. These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls are not intended for use with assays used to screen the blood supply. They are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HCV High Control
Detectable Analytes HCV
Type HCV Control
Level High
Packaging 5 x 1.2mL
Quantity 5 x 1.2 mL