Learn More
Thermo Scientific™ AcroMetrix™ HCV Controls
Catalog No. p-7000121
Change view
Click to view available options
Level:
High
Low
Mid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-279-125 Low
23-279-126 Mid
23-279-127 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-125 Supplier Thermo Scientific™ Supplier No. 963001
Only null left
Add to Cart
Add to Cart
Identify random or systematic errors in nucleic acid test procedures for hepatitis C viral RNA (HCV RNA) with these controls, which are for In Vitro Diagnostic Use.
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls. These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls are not intended for use with assays used to screen the blood supply. They are for In Vitro Diagnostic Use.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HCV Low Control
Detectable Analytes HCV
Type HCV Control
Level Low
Packaging 5 x 1.2mL
Quantity 5 x 1.2 mL