Learn More
Thermo Scientific™ AcroMetrix HCV Genotyping Panel (RUO)
Catalog No. 23279124 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 0.5 mL
Catalog No. Quantity
23-279-124 4 x 0.5 mL
1 options
Catalog No. 23-279-124 Supplier Thermo Scientific™ Supplier No. 942000
Add to Cart
Add to Cart
Assess the performance of nucleic acid test procedures that genotype HCV with the Thermo Scientific™ AcroMetrix™ HCV Genotyping Panel, which is for Research Use Only.
Assess the analytical performance of molecular test procedures designed to identify different genetic variants (genotypes) of human Hepatitis C Virus (HCV) with the Thermo Scientific™ AcroMetrix™ HCV Genotyping Panel (RUO). The panel is for Research Use Only and is not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description Genotypes 1, 2, 3 and 4
Detectable Analytes HCV
Type HCV Genotyping Panel
Quantity 4 x 0.5 mL