Learn More
Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO)
Shop All Thermo Scientific ProductsDescription
Assess the performance of nucleic acid test procedures for the quantitative and qualitative determination of Human Herpes Viruses (HHV) with the Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
Specifications
Specifications
| Description | AcroMetrix HSV-2 High CSF Control |
| Type | HSV-2 Control |
| Packaging | 5 x 0.5mL |
| Quantity | 5 x 0.5 mL |