Learn More
Description
Assess the performance of molecular diagnostic test procedures, including the RNA extraction step, for the quantitative and qualitative determination of human immunodeficiency virus type-1 (HIV-1) RNA with the Thermo Scientific™ AcroMetrix™ HIV-1 Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | AcroMetrix HIV-1 High Control |
| Detectable Analytes | HIV-1 |
| Type | HIV-1 Control |
| Packaging | 5 x 1.2mL |
| Quantity | 5 x 1.2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
