Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix Inhibition Panel
GSA_VA

Catalog No. 23279174 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
7 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-279-174 7 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-279-174 Supplier Thermo Scientific™ Supplier No. 956400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detect interfering substances that may inhibit nucleic acid amplification or serology assays with this panel, which is for Research Use Only.

Detect interfering substances that may inhibit nucleic acid amplification or serology assays with the Thermo Scientific™ AcroMetrix Inhibition Panel (RUO). Designed to mimic clinical conditions such as haemolysis, icterus and lipemia, this panel contains interfering substances found in serum, plasma or whole blood specimens. The panel is for Research Use Only and is not for use in diagnostic procedures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Inhibition Panel
Packaging 7 x 1mL
Quantity 7 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.