Learn More
Thermo Scientific™ AcroMetrix™ Multi-Analyte ctDNA Plasma Control A - E (RUO)
Shop All Thermo Scientific ProductsDescription
Validate and assess run-to-run assay variation of ctDNA somatic mutation panels with the Thermo Scientific™ AcroMetrix™ Multi-Analyte ctDNA Plasma Control A - E (RUO). The control is for research use only.
Validate and assess run-to-run assay variation of ctDNA somatic mutation panels. Intended for use with next generation sequencing (NGS), qualitative/quantitative PCR assays designed to identify somatic mutations in DNA from liquid biopsy samples. For research use only.
- Contains variants that cover different cancer types which include single nucleotide variant (SNV), INDEL (insertion, deletion), and Copy Number Variant (CNV)
- Kit contains one vial at specific variant allele frequency and CNV
- Full process control needs to go through extraction prior to library preparation or amplification
Specifications
Specifications
| Content And Storage | -20°C or lower |
| Description | AcroMetrix Multi-Analyte ctDNA Plasma Control A (RUO) |
| Detectable Analytes | Multiple |
| Type | Multi-Analyte ctDNA Control A |
| Packaging | Each kit contains 1 vial with 60 ng/mL DNA content in 2 mL plasma |
| Quantity | 1 Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.