Learn More
Thermo Scientific™ AcroMetrix™ Multi-Analyte ctDNA Plasma Control A - E (RUO)
Catalog No. 100957567 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Multi-Analyte ctDNA Control A
Multi-Analyte ctDNA Control B
Multi-Analyte ctDNA Control C
Multi-Analyte ctDNA Control D
Multi-Analyte ctDNA Control E
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-095-7567 Multi-Analyte ctDNA Control E
10-095-7563 Multi-Analyte ctDNA Control A
10-095-7564 Multi-Analyte ctDNA Control B
10-095-7565 Multi-Analyte ctDNA Control C
10-095-7566 Multi-Analyte ctDNA Control D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-095-7567 Supplier Thermo Scientific™ Supplier No. 957567
Add to Cart
Add to Cart
Validate and assess run-to-run assay variation of ctDNA somatic mutation panels with the Thermo Scientific™ AcroMetrix™ Multi-Analyte ctDNA Plasma Control A - E (RUO). The control is for research use only.
Validate and assess run-to-run assay variation of ctDNA somatic mutation panels. Intended for use with next generation sequencing (NGS), qualitative/quantitative PCR assays designed to identify somatic mutations in DNA from liquid biopsy samples. For research use only.
  • Contains variants that cover different cancer types which include single nucleotide variant (SNV), INDEL (insertion, deletion), and Copy Number Variant (CNV)
  • Kit contains one vial at specific variant allele frequency and CNV
  • Full process control needs to go through extraction prior to library preparation or amplification
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C or lower
Description AcroMetrix Multi-Analyte ctDNA Plasma Control E (RUO)
Detectable Analytes Multiple
Type Multi-Analyte ctDNA Control E
Packaging Each kit contains 1 vial with 60 ng/mL DNA content in 2 mL plasma
Quantity 1 Vial