Learn More
Thermo Scientific™ AcroMetrix™ MultiMix FFPE Control (RUO)
Catalog No. p-7000143
Change view
Click to view available options
Description:
MultiMix F
Target Gene:
FLT3, KIT, KRAS, EGFR, KDR, PIK3CA, HRAS, TP53, MET, SMO, FGFR1, FGFR3, NOTCH1, PDGFRA, SMARCB1, CSF1R
Catalog No. Description Target Gene
23-279-201 MultiMix F FLT3, KIT, KRAS, EGFR, KDR, PIK3CA, HRAS, TP53, MET, SMO, FGFR1, FGFR3, NOTCH1, PDGFRA, SMARCB1, CSF1R
1 options
Catalog No. 23-279-201 Supplier Thermo Scientific™ Supplier No. 957189
Assess performance of somatic genetic variant detection in research oncology testing with these controls, which are for Research Use Only.
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description MultiMix F
Type MultiMix FFPE Control
Packaging 5 samples
Target Gene FLT3, KIT, KRAS, EGFR, KDR, PIK3CA, HRAS, TP53, MET, SMO, FGFR1, FGFR3, NOTCH1, PDGFRA, SMARCB1, CSF1R