Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix Negative Control (IVD)
GSA_VA

Catalog No. 23279173 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-279-173 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-279-173 Supplier Thermo Scientific™ Supplier No. 962000A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Assess test procedures with a pre-screened negative plasma-based control, which is for In Vitro Diagnostic Use.

Assess test procedures with the Thermo Scientific™ AcroMetrix™ Negative Control. Used by clinical laboratories and diagnostic manufacturers, the AcroMetrix™ Negative Control is a well-characterized, pre-screened negative plasma-based control. The control is for In Vitro Diagnostic Use.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Negative Control
Packaging 5 x 1mL
Quantity 5 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.