Learn More
Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO)
GSA_VA
Catalog No. 23279190 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix PeliSpy Sero Type 17 Control
Detectable Analytes:
anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Catalog No. Description Detectable Analytes
23-279-190 AcroMetrix PeliSpy Sero Type 17 Control anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
1 options
Catalog No. 23-279-190 Supplier Thermo Scientific™ Supplier No. S2407
Monitor assays that test for HBsAg, anti-HBc, anti-HCV, anti-HIV-1/2 and anti-HTLV-1/2 with these controls, which are for Research Use Only.
Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix PeliSpy Sero Type 17 Control
Detectable Analytes anti-HCV, anti-HIV-1 and 2, anti-HTLV I and II, HBsAg
Sample Type Serum
Type Control
Packaging 10 x 5mL
Quantity 10 x 5 mL