Learn More
Description
Assess the analytical performance of molecular test procedures, including the nucleic acid extraction step, for the quantitative and qualitative determination of viruses related to transplant disease with the Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits, and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | AcroMetrix Adenovirus Plasma Panel |
| Type | Plasma Panel |
| Level | 1000, 10000, 100000, 1000000, 10000000 copies/mL |
| Packaging | 5 x 0.5mL |
| Quantity | 5 x 0.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
