Learn More
Thermo Scientific™ AcroMetrix™ VZV Controls (RUO)
GSA_VA
Catalog No. 23279161 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix VZV High CSF Control
AcroMetrix VZV High Plasma Control
AcroMetrix VZV Low CSF Control
Level:
High
Low
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level
23-279-161 AcroMetrix VZV High Plasma Control High
23-279-160 AcroMetrix VZV High CSF Control High
23-279-159 AcroMetrix VZV Low CSF Control Low
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-279-161 Supplier Thermo Scientific™ Supplier No. 954514
Add to Cart
Add to Cart
Identify increases in random or systematic error of nucleic acid test procedures for human VZV DNA with these controls, which are for Research Use Only.
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of human VZV DNA with the Thermo Scientific™ AcroMetrix™ VZV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix VZV High Plasma Control
Type VZV Control
Level High
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL