Learn More
Cytiva AcroPak™ 1500 Capsules with Supor EKV Membrane - 0.2 µm, 13 mm (1/2 in.) hose barb inlet/outlet connection, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12467
Encompass
For Cost-Effective Filtration of Buffers, Tissue Culture Media, and Other Biological Solutions-Newer technology recommended for simplified scale-up-Disposable design reduces labor costs associated with assembling, cleaning, and testing stainless steel filter holders-Encapsulated format for ease of use-Extended life with built-in MachV asymmetric prefilter layer for maximum flow and throughput performance-Low adsorption and high chemical compatibility with hydrophilic polyethersulfone membrane for high compatibility over the entire pH range and low protein binding-Gamma-irradiated, non-pyrogenic, and non-cytotoxic-Inherently hydrophilic for reliable integrity testing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.