Learn More
Cytiva AcroPak™ 20 Filters with Fluorodyne II Membrane, with Filling Bell - 0.2 µm, gamma-irradiated (3/pkg)
Catalog No. 502063194 Shop All Cytiva Products
Encompass
Efficient Processing of Liquid Volumes Typically Up To 20 L-Fluorodyne II membrane is compatible with aqueous and many organic solvents-AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations-AcroPak 200 Fluorodyne II capsules provide fast processing of clean batches-Double layer sterilizing membrane enhances reliability-Upstream air vent prevents vapor lock-Manufactured without the use of adhesives to minimize extractables-Pre-sterilized by gamma irradiation This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3194 Supplier Cytiva Supplier No. 12201
Efficient Processing of Liquid Volumes Typically Up To 20 L-Fluorodyne II membrane is compatible with aqueous and many organic solvents-AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations-AcroPak 200 Fluorodyne II capsules provide fast processing of clean batches-Double layer sterilizing membrane enhances reliability-Upstream air vent prevents vapor lock-Manufactured without the use of adhesives to minimize extractables-Pre-sterilized by gamma irradiation This product is not approved for use in medical, clinical, surgical or other patient applications.