Learn More
Cytiva AcroPak™ 300 Capsule with PTFE Membrane - 0.2 µm, stepped hose barb (3/pkg)
Catalog No. 502063178 Shop All Cytiva Products
Encompass
High Air Flow Rate Venting Applications-Offers broad chemical compatibility with PTFE membrane and polypropylene housing-100% integrity-tested to provide the high level of confidence you require This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3178 Supplier Cytiva Supplier No. 12082
High Air Flow Rate Venting Applications-Offers broad chemical compatibility with PTFE membrane and polypropylene housing-100% integrity-tested to provide the high level of confidence you require This product is not approved for use in medical, clinical, surgical or other patient applications.