Learn More
Cytiva AcroPak™ 400 Capsules with Fluorodyne II Membrane - 0.2 µm, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12478
Encompass_Preferred
Designed for Processing Volumes Up To 100 L-Fluorodyne II membrane is a hydrophilic polyvinylidene fluoride (PVDF) that offers higher flow rates with low protein and preservative binding, and broad chemical and temperature resistance-Suited for biological, pharmaceutical, and sterilizing filtration requirements-Scalable This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3203
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.