Learn More
Cytiva AcroPak™ 800 Capsules With Supor EKV Membrane - 0.2 m, 13 mm (1/2 in.) hose barb inlet/outlet connection with filling bell on outlet, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12464
Encompass_Preferred
For Buffers, Tissue Culture Media, and Other Biological Fluids-Newer technology recommended for simplified scale-up-High filtration area and compact size are ideal for upscale trials-Minimal hold-up volume-Good wettability for reliable integrity test-Sealed membrane and housing uses fusion technology to eliminate potential extractables-Gamma sterilized, non-pyrogenic, and provided with a removable filling bell (except sanitary flange option)-Hydrophilic polyethersulfone membrane for low adsorption and wide chemical compatibility-Very high flow rates and consistently higher total solution throughputs due to superior porosity over other membranes This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1885593
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.