Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 1000 Capsules with Supor Membrane - 0.2/0.2 m, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12996
Encompass_Preferred
Fast, Final Filtration and High Throughputs for Liquid Volumes Up To 150 LProvides higher throughputs and faster flow rates than similarly-sized competitive devicesIdeal for solutions requiring low protein bindingSaves money by increasing throughput with available built-in prefilter100% integrity testing assures sterile filtrate0.1 m Supor membrane version ensures sterile, mycoplasma-free cell culture mediaFusion-welded components eliminate the potential for release of extractables from sealing adhesivesSterilization by gamma irradiation eliminates potential contamination by EtO residuals This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.