Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration - 1 mL, 0.2 µm Supor membrane (5/pkg)
Catalog No. NC1001814 Shop All Cytiva Products
Encompass
Fast Processing with Efficient Removal of Particulates •New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume•New outlet tip geometry facilitates direct flow of samples into receiver plate without concerns of cross-contamination•Varied membrane and pore size selection offers efficient particulate removal•Manufactured in accordance with SBS guidelines, allowing plates to be run in manual, semi-automated and automated processes•Smooth top surface and textured window allow for easy labeling on the plates, as well as provide easy usage orientation through the A1 corner notch This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1001814 Supplier Cytiva Supplier No. 8119
Fast Processing with Efficient Removal of Particulates •New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume•New outlet tip geometry facilitates direct flow of samples into receiver plate without concerns of cross-contamination•Varied membrane and pore size selection offers efficient particulate removal•Manufactured in accordance with SBS guidelines, allowing plates to be run in manual, semi-automated and automated processes•Smooth top surface and textured window allow for easy labeling on the plates, as well as provide easy usage orientation through the A1 corner notch This product is not approved for use in medical, clinical, surgical or other patient applications.