Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Broth, Lennox

Catalog No. AC612725000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC612725000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC612725000 Supplier Thermo Scientific Chemicals Supplier No. 612725000
  • $187.40
    $147.65 / Each of 1

    Save $39.75

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Powder
Media Type LB Broth, Lennox
Product Type Microbiology Media
Quantity 500 g
What is the composition of LB Broth, Lennox (Cat. No. 612725000)?

LB Broth, Lennox (Cat. No. 612725000) is composed of 10.0 g/L casein digest peptone, 5.0 g/L sodium chloride, and 5.0 g/L yeast extract.

How should I prepare LB Broth, Lennox (Cat. No. 612725000)?

We recommend the following:
1. Mix 20 grams of the LB broth powder in 1 liter of purified water.
2. Agitate to completely dissolve.
3. Autoclave for 15 minutes at 121°C.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.