Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acrosin Monoclonal Antibody (ACR-2)
GREENER_CHOICE

Catalog No. 01675950
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-950 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-950 Supplier Invitrogen™ Supplier No. MA119180
  • $599.00
    $497.30 / Each of 1

    Save $101.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa. This antibody will not cross-react with human, canine or bovine. Immunocytochemistry: Membrane permeabilization (acetone) is essential.

ACR is the major proteinase present in the acrosome of mature spermatozoa. It is a typical serine proteinase with trypsin-like specificity. It is stored in the acrosome in its precursor form, proacrosin. The active enzyme functions in the lysis of the zona pellucida, thus facilitating penetration of the sperm through the innermost glycoprotein layers of the ovum. The mRNA for proacrosin is synthesized only in the postmeiotic stages of spermatogenesis. In humans proacrosin first appears in the haploid spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acrosin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ACR-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene ACR
Gene Accession No. P08001
Gene Alias 53 kDa fucose-binding protein; Acr; Acro; Acrosin; Acrosin heavy chain; acrosin light and heavy chain prepropeptide; Acrosin light chain; acrosin precursor (EC 3.4.21.10); acrosin prepropeptide; ACRS; preproacrosin; proacrosin
Gene Symbols ACR
Host Species Mouse
Immunogen Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397098
Target Species Pig
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.