Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acrylamide (40%) Solutions PROMO

Catalog No. J62480K2PM
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J62480K2PM 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J62480K2PM Supplier Thermo Scientific Chemicals Supplier No. J62480K2PR

Applications

  • Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins.
  • Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility

  • It is soluble in water.

Notes

  • Store in cool place.
  • Keep container tightly closed in a dry and well-ventilated place.
  • Light sensitive.
  • Keep away from acids, oxidizing agents, iron and iron salts, copper, brass, and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08 g/mol
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 1 L
UN Number UN3426
Beilstein 605349
Merck Index 14,129
Solubility Information It is Soluble in Water.
Chemical Name or Material Acrylamide, Bisacrylamide Free
Hazard Category H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Hazard Statement GHS H Statement: H340-H350-H372-H361-H302-H315-H319-H317. May cause genetic defects. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
EINECSNumber 201-173-7
TSCA Yes
Recommended Storage Keep Cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.