Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide (Flake-Like Crystals/Certified), ACS, Fisher Chemical

Catalog No. O1065500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O1065-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O1065-500

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym 2-Propenamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Solid
Boiling Point 125°C
Melting Point 82°C
Color White
pH 6.5 to 8.0
Quantity 500 g
Flash Point 40°C
Packaging Poly Bottle
Linear Formula CH2CHCONH2
UN Number 2074
Formula Weight 71.08
Grade Certified
Chemical Name or Material Acrylamide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Avoid contact with skin. Wash hands before breaks and immediately after handling the product. Avoid prolonged or repeated contact with skin. Do not taste or swallow. Do not take internally. . . . .

NFPA

Health: 2
Flammability:  2
Instability: 2
DOTInformation Class 6.1: Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.