Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide Solution (40%/Electrophoresis), Fisher BioReagents

Catalog No. BP14021 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1402-1 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP1402-1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
40% Acrylamide Solution is made from 3X recrystallized acrylamide and is suitable for electrophoretic separation of proteins and nucleic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1 , 7732-18-5
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Liquid
Acrylic Acid 0.003% max.
Boiling Point 125°C
Melting Point 83°C
Conductivity 5μmhos max.
Color Colorless
pH 6 to 7
Quantity 1 L
DNase DNase free
Packaging Amber Glass
Merck Index 15, 120
Grade Electrophoresis
Chemical Name or Material Acrylamide Solution, 40%
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Suspect cancer hazard. May cause heritable genetic damage. Toxic if swallowed. Harmful if absorbed through skin or if inhaled. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Hazardous polymerization may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood.

NFPA

Health:2
Flammability:2
Instability:2
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.