Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide, Ultrapure, Electrophoresis Grade, 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
79-06-1
79-06-1
CAS Max % 1
Packaging Poly Pail
Percent Purity 0.99
Grade Electrophoresis Grade
Melting Point 83° to 86°C
pH 5.5 to 7.0
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.