Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide, Ultrapure, Electrophoresis Grade, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608438 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-438 100 g Fiber Drum
18-608-439 500 g Fiber Drum
18-608-440 2.5 kg Fiber Drum
18-608-441 12 kg Fiber Drum
18-608-437 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-438 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1029100GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79-06-1

Specifications

Physical Form Solid
Melting Point 83° to 86°C
CAS Max % 1
pH 5.5 to 7.0
Quantity 100 g
Packaging Fiber Drum
Percent Purity 99%
Grade Electrophoresis
Chemical Name or Material Acrylamide
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.