Learn More
Eisco™ Acrylic Block Set with Wooden Storage Case, Set of 7
SCI_ED
Catalog No. S27414 Shop All Eisco Products
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S27414 Acrylic
1 options
Catalog No. S27414 Supplier Eisco™ Supplier No. PH0572A
Add to Cart
Add to Cart
Optical acrylic block set with storage box includes 7 different acrylic blocks. Designed for demonstrations and experiments in optics, refraction, spectrometry, and wavelengths.
  • Set includes one of each: rectangle, semi-circle, bi-convex, bi-concave and three triangular blocks of varying sizes
  • Each block is 15 mm (0.6 in.) thick
  • Includes wooden storage case with latch
  • Designed for experiments and demonstrations in optics, light refraction, spectrometry, and wavelengths
  • Also used for photography lighting and distortion effects
Specifications
Material Acrylic
Includes One of Each Shape: Rectangle, Semi-Circle, Bi-Convex, Bi-Concave and 3 Triangular Blocks of Varying Sizes
For Use With (Application) Experiments and Demonstrations in Optics, Light Refraction, Spectrometry, and Wavelength
Product Type Block Set
Shape Various